The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000255.3 | NP_000246.2 | 6 - 7 | 1593 | 117 |
XM_005249143.3 | XP_005249200.1 | 6 - 7 | 1498 | 117 | |
GenBank mRNA | AB208845.1 | - | 1 - 2 | 171 | 117 |
AK292568.1 | - | 6 - 7 | 1569 | 117 | |
AK312611.1 | - | 6 - 7 | 1568 | 117 | |
BC016282.1 | - | 6 - 7 | 1456 | 117 | |
BT007434.1 | - | 5 - 6 | 1328 | 117 | |
BX647789.1 | - | 6 - 7 | 1418 | 117 | |
KF030882.1 | - | 5 - 6 | 1328 | 117 | |
KU178201.1 | - | 5 - 6 | 1328 | 117 | |
KU178202.1 | - | 5 - 6 | 1328 | 117 | |
M65131.1 | - | 6 - 7 | 1404 | 117 |