Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
Chromosome Location
Chr.14: 55148116 - 55191678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146015.1 NP_001139487.1 14 - 15 2088 152
NM_014750.4 NP_055565.3 14 - 15 2088 152
XM_017021840.1 XP_016877329.1 14 - 15 1980 152
GenBank mRNA AB076695.1 - 14 - 15 2088 152
AK291847.1 - 14 - 15 1965 152
AK297480.1 - 13 - 14 1841 152
AK299338.1 - 14 - 15 1965 152
BC010658.1 - 14 - 15 1980 152
BC016276.2 - 13 - 14 1874 152
BT007344.1 - 12 - 13 1628 152
BX248255.1 - 14 - 15 1954 152
D13633.3 - 14 - 15 1961 152

View Full Product Details