Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
Chromosome Location
Chr.14: 55148116 - 55191678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146015.1 NP_001139487.1 3 - 4 656 78
NM_014750.4 NP_055565.3 3 - 4 656 78
XM_017021840.1 XP_016877329.1 3 - 4 548 78
GenBank mRNA AB076695.1 - 3 - 4 656 78
AK291847.1 - 3 - 4 533 78
AK299338.1 - 3 - 4 533 78
BC010658.1 - 3 - 4 548 78
BC016276.2 - 2 - 3 442 78
BT007344.1 - 1 - 2 196 78
BX248255.1 - 3 - 4 522 78
D13633.3 - 3 - 4 529 78

View Full Product Details