The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182509.3 | NP_872315.2 | 2 - 3 | 465 | 63 |
NM_199296.2 | NP_954993.1 | 2 - 3 | 465 | 63 | |
XM_011536489.1 | XP_011534791.1 | 2 - 3 | 355 | 63 | |
GenBank mRNA | AJ583024.1 | - | 2 - 3 | 426 | 63 |
AK056709.1 | - | 3 - 4 | 519 | 63 | |
AK075445.1 | - | 2 - 3 | 448 | 63 | |
AK291600.1 | - | 2 - 3 | 414 | 63 | |
BC033140.1 | - | 1 - 2 | 63 | ||
BC101019.1 | - | 2 - 3 | 414 | 63 | |
BC101020.1 | - | 2 - 3 | 414 | 63 | |
BC101021.2 | - | 2 - 3 | 414 | 63 | |
BC101022.2 | - | 2 - 3 | 414 | 63 | |
BX248277.1 | - | 2 - 3 | 398 | 63 | |
BX248770.1 | - | 2 - 3 | 404 | 63 | |
BX248771.1 | - | 2 - 3 | 785 | 63 | |
CR749863.1 | - | 2 - 3 | 414 | 63 | |
DB025298.1 | - | 2 - 3 | 466 | 63 |