The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003082.3 | NP_003073.1 | 2 - 3 | 395 | 153 |
GenBank mRNA | AK314933.1 | - | 2 - 3 | 395 | 153 |
BC014984.1 | - | 2 - 3 | 303 | 153 | |
BC019038.2 | - | 2 - 3 | 299 | 153 | |
U44754.1 | - | 2 - 3 | 303 | 153 | |
Z47542.1 | - | 2 - 3 | 315 | 153 |