The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015562.1 | NP_056377.1 | 10 - 11 | 1383 | 95 |
XM_011512670.2 | XP_011510972.1 | 10 - 11 | 1331 | 95 | |
XM_011512671.2 | XP_011510973.1 | 9 - 10 | 1188 | 95 | |
GenBank mRNA | AB018337.1 | - | 10 - 11 | 1311 | 95 |
AK303630.1 | - | 9 - 10 | 1188 | 95 | |
AK307899.1 | - | 6 - 7 | 944 | 95 | |
BC028986.1 | - | 9 - 10 | 1215 | 95 | |
BC047420.1 | - | 10 - 11 | 1333 | 95 |