Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
Chromosome Location
Chr.6: 485138 - 693141 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018303.5 NP_060773.3 25 - 26 2724 70
NR_073064.1 27 - 28 2886 70
XM_017011018.1 XP_016866507.1 26 - 27 2770 70
XM_017011019.1 XP_016866508.1 26 - 27 2932 70
XM_017011020.1 XP_016866509.1 26 - 27 2792 70
XM_017011021.1 XP_016866510.1 25 - 26 3028 70
XM_017011022.1 XP_016866511.1 25 - 26 2724 70
XM_017011023.1 XP_016866512.1 25 - 26 2724 70
XM_017011024.1 XP_016866513.1 26 - 27 2770 70
GenBank mRNA AJ414403.1 - 25 - 26 2648 70
AJ420556.1 - 3 - 4 243 70
AK001888.1 - 21 - 22 2139 70
AK056564.1 - 25 - 26 2692 70
AK302941.1 - 15 - 16 1465 70
AK314628.1 - 25 - 26 2724 70
AL833209.1 - 23 - 24 2280 70
AL833213.1 - 25 - 26 2670 70
BC016918.1 - 22 - 23 2204 70
BC080542.1 - 27 - 28 2856 70

View Full Product Details