Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
Chromosome Location
Chr.6: 485138 - 693141 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018303.5 NP_060773.3 6 - 7 830 70
NR_073064.1 8 - 9 992 70
XM_017011018.1 XP_016866507.1 7 - 8 876 70
XM_017011019.1 XP_016866508.1 7 - 8 1038 70
XM_017011020.1 XP_016866509.1 7 - 8 898 70
XM_017011021.1 XP_016866510.1 6 - 7 1134 70
XM_017011022.1 XP_016866511.1 6 - 7 830 70
XM_017011023.1 XP_016866512.1 6 - 7 830 70
XM_017011024.1 XP_016866513.1 7 - 8 876 70
XM_017011025.1 XP_016866514.1 6 - 7 830 70
XM_017011026.1 XP_016866515.1 6 - 7 830 70
GenBank mRNA AJ414403.1 - 6 - 7 754 70
AK001888.1 - 2 - 3 245 70
AK056564.1 - 6 - 7 798 70
AK314628.1 - 6 - 7 830 70
AL833209.1 - 4 - 5 386 70
AL833213.1 - 6 - 7 776 70
BC016918.1 - 3 - 4 310 70
BC080542.1 - 8 - 9 962 70

View Full Product Details