The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018638.4 | NP_061108.2 | 5 - 6 | 1072 | 113 |
XM_017019580.1 | XP_016875069.1 | 5 - 6 | 1069 | 113 | |
GenBank mRNA | AF207600.2 | - | 5 - 6 | 1138 | 113 |
AK074425.1 | - | 5 - 6 | 1017 | 113 | |
AL137750.1 | - | 5 - 6 | 1021 | 113 | |
BC066907.1 | - | 5 - 6 | 1072 | 113 |