The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005482.2 | NP_005473.1 | 10 - 11 | 1119 | 112 |
GenBank mRNA | AF022913.1 | - | 10 - 11 | 1091 | 112 |
AK304340.1 | - | 8 - 9 | 874 | 112 | |
AK313015.1 | - | 10 - 11 | 1102 | 112 | |
BC020737.1 | - | 10 - 11 | 1098 | 112 | |
BX648473.1 | - | 7 - 8 | 821 | 112 | |
Y07596.1 | - | 10 - 11 | 1078 | 112 |