Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGK
Entrez Gene ID
10026
Gene Name
phosphatidylinositol glycan anchor biosynthesis class K
Gene Aliases
GPI8
Chromosome Location
Chr.1: 77088981 - 77219447 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005482.2 NP_005473.1 3 - 4 282 94
GenBank mRNA AF022913.1 - 3 - 4 254 94
AK313015.1 - 3 - 4 265 94
BC020737.1 - 3 - 4 261 94
DA263434.1 - 3 - 4 282 94
Y07596.1 - 3 - 4 241 94

View Full Product Details