The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302777.1 | NP_001289706.1 | 6 - 7 | 1255 | 104 |
NM_004332.3 | NP_004323.2 | 5 - 6 | 1027 | 104 | |
GenBank mRNA | AJ617684.1 | - | 5 - 6 | 675 | 104 |
AK314909.1 | - | 6 - 7 | 909 | 104 | |
BC106901.1 | - | 5 - 6 | 742 | 104 | |
X57653.1 | - | 1 - 2 | 119 | 104 | |
X81372.1 | - | 6 - 7 | 823 | 104 |