The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000379.3 | NP_000370.2 | 17 - 18 | 1930 | 119 |
XM_011533095.2 | XP_011531397.1 | 17 - 18 | 1959 | 119 | |
XM_011533096.2 | XP_011531398.1 | 17 - 18 | 1961 | 119 | |
GenBank mRNA | D11456.2 | - | 17 - 18 | 1908 | 119 |
U06117.1 | - | 17 - 18 | 1914 | 119 | |
U39487.1 | - | 17 - 18 | 1885 | 119 |