The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193513.1 | NP_001180442.1 | 2 - 3 | 152 | 100 |
NM_001193514.1 | NP_001180443.1 | 2 - 3 | 152 | 100 | |
NM_017964.3 | NP_060434.2 | 2 - 3 | 152 | 100 | |
XM_011532959.2 | XP_011531261.1 | 2 - 3 | 129 | 100 | |
XM_011532960.2 | XP_011531262.1 | 2 - 3 | 408 | 100 | |
XM_011532961.2 | XP_011531263.1 | 2 - 3 | 129 | 100 | |
XM_011532963.2 | XP_011531265.1 | 2 - 3 | 129 | 100 | |
XM_017004463.1 | XP_016859952.1 | 2 - 3 | 129 | 100 | |
XM_017004464.1 | XP_016859953.1 | 2 - 3 | 129 | 100 | |
XM_017004465.1 | XP_016859954.1 | 2 - 3 | 129 | 100 | |
XM_017004466.1 | XP_016859955.1 | 2 - 3 | 144 | 100 | |
GenBank mRNA | AF173387.1 | - | - | 104 | 100 |
AK055663.1 | - | 2 - 3 | 129 | 100 | |
AK096672.1 | - | 2 - 3 | 129 | 100 | |
BC066903.1 | - | 2 - 3 | 129 | 100 | |
EF560719.1 | - | 2 - 3 | 130 | 100 | |
EF560726.1 | - | 2 - 3 | 130 | 100 |