Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPATCH2
Entrez Gene ID
55105
Gene Name
G-patch domain containing 2
Gene Aliases
CT110, GPATC2, PPP1R30
Chromosome Location
Chr.1: 217426992 - 217631312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297754.1 NP_001284683.1 4 - 5 1146 66
NM_018040.3 NP_060510.1 4 - 5 1146 66
XM_011509689.2 XP_011507991.1 4 - 5 1356 66
XM_011509690.2 XP_011507992.1 4 - 5 1356 66
XM_011509693.2 XP_011507995.1 4 - 5 1356 66
XM_011509694.2 XP_011507996.1 4 - 5 1356 66
XM_017001592.1 XP_016857081.1 4 - 5 1356 66
XM_017001593.1 XP_016857082.1 4 - 5 1356 66
GenBank mRNA AK001114.1 - 4 - 5 1113 66
AK291238.1 - 4 - 5 1111 66
BC042193.1 - 4 - 5 1063 66
BC063474.1 - 4 - 5 1095 66

View Full Product Details