The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025128.4 | NP_079404.3 | 7 - 8 | 1105 | 107 |
XM_005274307.2 | XP_005274364.1 | 7 - 8 | 1103 | 107 | |
XM_011545269.1 | XP_011543571.1 | 8 - 9 | 920 | 107 | |
XM_011545270.1 | XP_011543572.1 | 8 - 9 | 920 | 107 | |
GenBank mRNA | AF425646.1 | - | 7 - 8 | 752 | 107 |
AK024665.1 | - | 7 - 8 | 690 | 107 | |
AK095326.1 | - | 5 - 6 | 1146 | 107 | |
AK126820.1 | - | 8 - 9 | 1071 | 107 | |
AK223564.1 | - | 7 - 8 | 1064 | 104 | |
AL353934.1 | - | 1 - 2 | 135 | 107 | |
BC009999.2 | - | 7 - 8 | 1028 | 107 |