Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGM1
Entrez Gene ID
5236
Gene Name
phosphoglucomutase 1
Gene Aliases
CDG1T, GSD14
Chromosome Location
Chr.1: 63593276 - 63660245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172818.1 NP_001166289.1 3 - 4 858 108
NM_001172819.1 NP_001166290.1 3 - 4 649 108
NM_002633.2 NP_002624.2 3 - 4 772 108
GenBank mRNA AK293298.1 - 3 - 4 649 108
AK294186.1 - 4 - 5 731 108
AK298505.1 - 3 - 4 579 108
AK300100.1 - 3 - 4 844 108
AK312254.1 - 3 - 4 637 108
BC001756.2 - 3 - 4 685 108
BC019920.1 - 3 - 4 772 108
BC067763.1 - 3 - 4 650 108
BC090856.1 - 3 - 4 617 108
BT006961.1 - 3 - 4 559 108
M83088.1 - 3 - 4 621 108

View Full Product Details