The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005255867.1 | XP_005255924.1 | 18 - 19 | 2910 | 66 |
XM_005255868.1 | XP_005255925.1 | 19 - 20 | 2922 | 66 | |
XM_011522970.1 | XP_011521272.1 | 19 - 20 | 2922 | 66 | |
XM_011522971.2 | XP_011521273.1 | 19 - 20 | 2922 | 66 | |
XM_011522973.2 | XP_011521275.1 | 19 - 20 | 2922 | 66 | |
XM_017023094.1 | XP_016878583.1 | 19 - 20 | 2922 | 66 | |
XM_017023095.1 | XP_016878584.1 | 19 - 20 | 2922 | 66 | |
XM_017023096.1 | XP_016878585.1 | 19 - 20 | 2922 | 66 | |
XM_017023098.1 | XP_016878587.1 | 5 - 6 | 1885 | 66 | |
XM_017023099.1 | XP_016878588.1 | 5 - 6 | 1251 | 66 | |
GenBank mRNA | AB023222.1 | - | 14 - 15 | 2339 | 66 |
AK026552.1 | - | 4 - 5 | 869 | 66 | |
AK304790.1 | - | 18 - 19 | 2972 | 66 | |
AK307929.1 | - | 18 - 19 | 2930 | 66 | |
AK308390.1 | - | 18 - 19 | 2920 | 66 | |
BC136433.1 | - | 18 - 19 | 2915 | 66 | |
CR749645.1 | - | 18 - 19 | 2929 | 66 |