Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPGRIP1L
Entrez Gene ID
23322
Gene Name
RPGRIP1 like
Gene Aliases
CORS3, FTM, JBTS7, MKS5, NPHP8, PPP1R134
Chromosome Location
Chr.16: 53599239 - 53703934 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005255867.1 XP_005255924.1 6 - 7 809 96
XM_005255868.1 XP_005255925.1 6 - 7 809 96
XM_011522970.1 XP_011521272.1 6 - 7 809 96
XM_011522971.2 XP_011521273.1 6 - 7 809 96
XM_011522973.2 XP_011521275.1 6 - 7 809 96
XM_017023094.1 XP_016878583.1 6 - 7 809 96
XM_017023095.1 XP_016878584.1 6 - 7 809 96
XM_017023096.1 XP_016878585.1 6 - 7 809 96
XM_017023097.1 XP_016878586.1 6 - 7 809 96
XM_017023100.1 XP_016878589.1 6 - 7 809 96
GenBank mRNA AB023222.1 - 2 - 3 238 96
AK096333.1 - 6 - 7 820 96
AK304790.1 - 6 - 7 871 96
AK308390.1 - 6 - 7 820 96
BC017977.1 - 6 - 7 822 96
BC136433.1 - 6 - 7 814 96
CR749645.1 - 6 - 7 828 96

View Full Product Details