Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIAS4
Entrez Gene ID
51588
Gene Name
protein inhibitor of activated STAT 4
Gene Aliases
PIAS-gamma, PIASY, Piasg, ZMIZ6
Chromosome Location
Chr.19: 4007598 - 4039386 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015897.3 NP_056981.2 2 - 3 622 88
XM_011528060.2 XP_011526362.1 2 - 3 965 88
XM_017026868.1 XP_016882357.1 1 - 2 432 88
GenBank mRNA AF077952.1 - 2 - 3 459 88
AF164437.1 - 1 - 2 432 88
AK222726.1 - 2 - 3 462 88
AK315530.1 - 2 - 3 502 88
BC010047.2 - 2 - 3 449 88
BC029874.1 - 2 - 3 469 88
BC066895.1 - 2 - 3 472 88

View Full Product Details