Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286164.1 NP_001273093.1 15 - 16 2713 92
NM_004319.2 NP_004310.1 15 - 16 2713 92
NM_207108.2 NP_996991.1 15 - 16 2713 92
XM_017001340.1 XP_016856829.1 15 - 16 2687 92
XM_017001341.1 XP_016856830.1 15 - 16 2687 92
GenBank mRNA AB006627.1 - 15 - 16 2511 92
AK295126.1 - 15 - 16 2665 92
BC015438.1 - 3 - 4 357 92
BC142697.1 - 15 - 16 2492 92
BC146795.1 - 15 - 16 2511 92

View Full Product Details