Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286164.1 NP_001273093.1 16 - 17 2902 76
NM_004319.2 NP_004310.1 16 - 17 2902 76
NM_207108.2 NP_996991.1 16 - 17 2902 76
XM_017001340.1 XP_016856829.1 16 - 17 2876 76
XM_017001341.1 XP_016856830.1 16 - 17 2876 76
GenBank mRNA AB006627.1 - 16 - 17 2700 76
AK295126.1 - 16 - 17 2854 76
BC015438.1 - 4 - 5 546 76
BC142697.1 - 16 - 17 2681 76
BC146795.1 - 16 - 17 2700 76

View Full Product Details