Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286164.1 NP_001273093.1 18 - 19 3307 87
NM_004319.2 NP_004310.1 18 - 19 3307 87
NM_207108.2 NP_996991.1 18 - 19 3307 87
XM_017001340.1 XP_016856829.1 18 - 19 3281 87
XM_017001341.1 XP_016856830.1 18 - 19 3281 87
GenBank mRNA AB006627.1 - 18 - 19 3105 87
AK295126.1 - 18 - 19 3259 87
BC015438.1 - 6 - 7 951 87
BC142697.1 - 18 - 19 3086 87
BC146795.1 - 18 - 19 3105 87

View Full Product Details