The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286164.1 | NP_001273093.1 | 20 - 21 | 3595 | 61 |
NM_004319.2 | NP_004310.1 | 20 - 21 | 3595 | 61 | |
NM_207108.2 | NP_996991.1 | 20 - 21 | 3595 | 61 | |
XM_017001340.1 | XP_016856829.1 | 20 - 21 | 3569 | 61 | |
XM_017001341.1 | XP_016856830.1 | 20 - 21 | 3569 | 61 | |
GenBank mRNA | AB006627.1 | - | 20 - 21 | 3393 | 61 |
AK295126.1 | - | 20 - 21 | 3547 | 61 | |
BC015438.1 | - | 8 - 9 | 1239 | 61 | |
BC142697.1 | - | 20 - 21 | 3374 | 61 | |
BC146795.1 | - | 20 - 21 | 3393 | 61 |