Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004319.2 NP_004310.1 22 - 23 3877 60
XM_017001340.1 XP_016856829.1 22 - 23 3851 60
GenBank mRNA AB006627.1 - 22 - 23 3675 60
BC146795.1 - 22 - 23 3675 60

View Full Product Details