The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013381.2 | NP_037513.1 | 4 - 5 | 1363 | 82 |
XM_005268819.4 | XP_005268876.1 | 4 - 5 | 1468 | 82 | |
XM_011538248.2 | XP_011536550.1 | 2 - 3 | 1541 | 82 | |
XM_017019243.1 | XP_016874732.1 | 4 - 5 | 1468 | 82 | |
XM_017019244.1 | XP_016874733.1 | 5 - 6 | 811 | 82 | |
GenBank mRNA | AF126372.1 | - | 4 - 5 | 1429 | 82 |
AY358765.1 | - | 4 - 5 | 1356 | 82 | |
BC142706.1 | - | 4 - 5 | 1363 | 82 | |
BC150181.1 | - | 4 - 5 | 1363 | 82 |