The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079515.2 | NP_001072983.1 | 5 - 6 | 676 | 134 |
NM_001287801.1 | NP_001274730.1 | 5 - 6 | 628 | 134 | |
NM_001287802.1 | NP_001274731.1 | 4 - 5 | 599 | 134 | |
NM_003193.4 | NP_003184.1 | 5 - 6 | 628 | 134 | |
GenBank mRNA | AK093397.1 | - | 4 - 5 | 504 | 134 |
AK292287.1 | - | 5 - 6 | 533 | 134 | |
AK296185.1 | - | 5 - 6 | 558 | 134 | |
AK298050.1 | - | 5 - 6 | 614 | 134 | |
BC008654.1 | - | 5 - 6 | 520 | 134 | |
BT007086.1 | - | 4 - 5 | 452 | 134 | |
U61232.1 | - | 5 - 6 | 581 | 134 |