Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE2
Entrez Gene ID
221184
Gene Name
copine 2
Gene Aliases
COPN2, CPN2
Chromosome Location
Chr.16: 57092543 - 57147966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152727.5 NP_689940.3 12 - 13 1402 109
GenBank mRNA AF492484.1 - 12 - 13 1353 109
AK292269.1 - 11 - 12 1311 109
AK315394.1 - 12 - 13 1305 109
BC053658.1 - 13 - 14 1476 109
CR749617.1 - 11 - 12 1063 109

View Full Product Details