The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032206.4 | NP_115582.4 | 2 - 3 | 422 | 93 |
XM_005256193.2 | XP_005256250.1 | 3 - 4 | 483 | 93 | |
XM_005256194.3 | XP_005256251.1 | 3 - 4 | 694 | 93 | |
XM_005256195.2 | XP_005256252.1 | 3 - 4 | 483 | 93 | |
XM_005256196.2 | XP_005256253.1 | 3 - 4 | 483 | 93 | |
XM_005256197.2 | XP_005256254.1 | 3 - 4 | 483 | 93 | |
XM_005256199.3 | XP_005256256.1 | 3 - 4 | 483 | 93 | |
XM_005256201.3 | XP_005256258.1 | 3 - 4 | 483 | 93 | |
XM_006721297.2 | XP_006721360.1 | 3 - 4 | 471 | 93 | |
XM_006721298.2 | XP_006721361.1 | 3 - 4 | 483 | 93 | |
XM_006721300.2 | XP_006721363.1 | 3 - 4 | 483 | 93 | |
XM_011523373.2 | XP_011521675.1 | 3 - 4 | 630 | 93 | |
XM_011523375.2 | XP_011521677.1 | 3 - 4 | 483 | 93 | |
XM_017023766.1 | XP_016879255.1 | 3 - 4 | 483 | 93 | |
XM_017023767.1 | XP_016879256.1 | 3 - 4 | 471 | 93 | |
XM_017023768.1 | XP_016879257.1 | 3 - 4 | 483 | 93 | |
XM_017023769.1 | XP_016879258.1 | 3 - 4 | 471 | 93 | |
XM_017023770.1 | XP_016879259.1 | 3 - 4 | 483 | 93 | |
XM_017023771.1 | XP_016879260.1 | 3 - 4 | 483 | 93 | |
GenBank mRNA | AK074182.1 | - | 2 - 3 | 435 | 93 |
AK097030.1 | - | 3 - 4 | 472 | 93 | |
DB293035.1 | - | 3 - 4 | 555 | 93 |