The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007006.2 | NP_008937.1 | 2 - 3 | 466 | 103 |
GenBank mRNA | AJ001810.1 | - | 2 - 3 | 319 | 103 |
AK312721.1 | - | 2 - 3 | 469 | 103 | |
AL535307.3 | - | 2 - 3 | 466 | 103 | |
BC001403.2 | - | 2 - 3 | 427 | 103 | |
BX537360.1 | - | 2 - 3 | 470 | 103 | |
CR456919.1 | - | 2 - 3 | 319 | 103 |