The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258392.2 | NP_001245321.1 | 4 - 5 | 849 | 61 |
NM_001258393.2 | NP_001245322.1 | 3 - 4 | 762 | 61 | |
GenBank mRNA | AK302006.1 | - | 3 - 4 | 605 | 61 |
AK302158.1 | - | 3 - 4 | 680 | 61 | |
BU848809.1 | - | 4 - 5 | 695 | 61 |