The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008707.1 | NP_001008707.1 | 10 - 11 | 1137 | 63 |
NM_004434.2 | NP_004425.2 | 9 - 10 | 1080 | 63 | |
XM_005267397.1 | XP_005267454.1 | 10 - 11 | 1207 | 63 | |
XM_005267398.3 | XP_005267455.1 | 10 - 11 | 1140 | 63 | |
XM_005267399.4 | XP_005267456.2 | 10 - 11 | 1179 | 63 | |
XM_005267400.4 | XP_005267457.1 | 9 - 10 | 1106 | 63 | |
XM_011536540.2 | XP_011534842.1 | 9 - 10 | 1153 | 63 | |
XM_011536541.1 | XP_011534843.1 | 10 - 11 | 1220 | 63 | |
XM_011536542.2 | XP_011534844.1 | 10 - 11 | 1179 | 63 | |
XM_017021074.1 | XP_016876563.1 | 9 - 10 | 1150 | 63 | |
GenBank mRNA | AK126980.1 | - | 11 - 12 | 1093 | 63 |
AK299812.1 | - | 10 - 11 | 1416 | 63 | |
AK307925.1 | - | 9 - 10 | 1072 | 63 | |
BC032541.1 | - | 9 - 10 | 1056 | 63 | |
BC033043.1 | - | 9 - 10 | 1080 | 63 | |
BX247979.1 | - | 10 - 11 | 1149 | 63 | |
U97018.1 | - | 9 - 10 | 1079 | 63 |