The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017000537.1 | XP_016856026.1 | 1 - 2 | 258 | 127 |
GenBank mRNA | BC037166.2 | - | 1 - 2 | 204 | 127 |
CB991812.1 | - | 1 - 2 | 268 | 127 | |
HY005615.1 | - | 1 - 2 | 264 | 127 | |
M59371.1 | - | 1 - 2 | 199 | 127 |