The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242339.1 | NP_001229268.1 | 10 - 11 | 1069 | 78 |
NM_001323067.1 | NP_001309996.1 | 8 - 9 | 808 | 78 | |
NM_001323068.1 | NP_001309997.1 | 8 - 9 | 945 | 78 | |
NM_001323069.1 | NP_001309998.1 | 8 - 9 | 940 | 78 | |
NM_001323070.1 | NP_001309999.1 | 3 - 4 | 755 | 78 | |
NM_001323071.1 | NP_001310000.1 | 9 - 10 | 1061 | 78 | |
NM_001323072.1 | NP_001310001.1 | 9 - 10 | 1168 | 78 | |
NM_001323073.1 | NP_001310002.1 | 4 - 5 | 429 | 78 | |
NM_001323074.1 | NP_001310003.1 | 3 - 4 | 755 | 78 | |
NM_002627.4 | NP_002618.1 | 8 - 9 | 945 | 78 | |
XM_005252465.4 | XP_005252522.1 | 8 - 9 | 990 | 78 | |
XM_005252466.4 | XP_005252523.1 | 8 - 9 | 990 | 78 | |
XM_005252467.2 | XP_005252524.1 | 8 - 9 | 914 | 78 | |
XM_006717449.1 | XP_006717512.1 | 8 - 9 | 914 | 78 | |
GenBank mRNA | AK092597.1 | - | 10 - 11 | 1069 | 78 |
AK291841.1 | - | 8 - 9 | 917 | 78 | |
AK308003.1 | - | 8 - 9 | 908 | 78 | |
BC002536.1 | - | 8 - 9 | 905 | 78 | |
BC029138.1 | - | 8 - 9 | 945 | 78 | |
D25328.1 | - | 8 - 9 | 902 | 78 |