The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002864.2 | NP_002855.2 | 34 - 35 | 4406 | 78 |
XM_017019764.1 | XP_016875253.1 | 34 - 35 | 4405 | 78 | |
XM_017019766.1 | XP_016875255.1 | 31 - 32 | 4305 | 78 | |
GenBank mRNA | AK313639.1 | - | 34 - 35 | 4405 | 78 |
BC111756.1 | - | 31 - 32 | 4305 | 78 | |
X54380.1 | - | 34 - 35 | 4406 | 78 |