The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020408.5 | NP_065141.3 | 2 - 3 | 418 | 138 |
GenBank mRNA | AF088060.1 | - | 2 - 3 | 198 | 138 |
AF170070.1 | - | 2 - 3 | 214 | 138 | |
AF285118.1 | - | 2 - 3 | 214 | 138 | |
AK291158.1 | - | 2 - 3 | 210 | 138 | |
BC009552.2 | - | 2 - 3 | 269 | 138 | |
BI758258.1 | - | 2 - 3 | 407 | 138 | |
HY168919.1 | - | 2 - 3 | 419 | 138 |