Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR1
Entrez Gene ID
2825
Gene Name
G protein-coupled receptor 1
Gene Aliases
-
Chromosome Location
Chr.2: 206175316 - 206218047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098199.1 NP_001091669.1 1 - 2 233 61
GenBank mRNA CB269800.1 - 1 - 2 32 61

View Full Product Details