Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
Chromosome Location
Chr.1: 203216079 - 203229732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270509.1 NP_001257438.1 4 - 5 412 86
NM_003465.2 NP_003456.1 4 - 5 412 86
NR_045784.1 4 - 5 412 86
NR_045785.1 4 - 5 412 86
GenBank mRNA AK131093.1 - 6 - 7 4672 86
AK225245.1 - 4 - 5 352 86
AK309341.1 - 4 - 5 86
AK314331.1 - 4 - 5 86
BC069614.1 - 3 - 4 281 86
BC103695.1 - 4 - 5 86
BC105680.1 - 4 - 5 86
BC105681.1 - 4 - 5 86
BC105682.1 - 4 - 5 328 86
BP304256.1 - 4 - 5 412 86
U29615.1 - 4 - 5 328 86
U62662.1 - 4 - 5 325 86

View Full Product Details