Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
Chromosome Location
Chr.1: 203216079 - 203229732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256125.1 NP_001243054.2 4 - 5 516 59
NM_003465.2 NP_003456.1 5 - 6 573 59
NR_045784.1 5 - 6 573 59
NR_045785.1 5 - 6 573 59
GenBank mRNA AK055165.1 - 4 - 5 456 59
AK309341.1 - 5 - 6 515 59
AK314331.1 - 5 - 6 512 59
BC069614.1 - 4 - 5 442 59
BC103695.1 - 5 - 6 489 59
BC105680.1 - 5 - 6 489 59
BC105681.1 - 5 - 6 489 59
BC105682.1 - 5 - 6 489 59
U29615.1 - 5 - 6 489 59
U62662.1 - 5 - 6 486 59

View Full Product Details