Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST3H3
Entrez Gene ID
8290
Gene Name
histone cluster 3, H3
Gene Aliases
H3.4, H3/g, H3FT, H3t
Chromosome Location
Chr.1: 228424845 - 228425325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003493.2 NP_003484.1 1 - 1 444 76
GenBank mRNA BC069079.1 - 1 - 1 444 76
BC101837.1 - 1 - 1 492 76
BC101839.1 - 1 - 1 492 76

View Full Product Details