The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052962.2 | NP_443194.1 | 6 - 7 | 945 | 168 |
NM_181309.1 | NP_851826.1 | 5 - 6 | 849 | 168 | |
GenBank mRNA | AJ297262.1 | - | 4 - 5 | 569 | 168 |
AJ313161.1 | - | 5 - 6 | 851 | 168 | |
AJ313162.1 | - | 6 - 7 | 947 | 168 | |
AY040566.1 | - | 5 - 6 | 822 | 168 | |
AY040567.1 | - | 6 - 7 | 918 | 168 | |
AY044429.1 | - | 5 - 6 | 784 | 168 | |
AY358111.1 | - | 5 - 6 | 847 | 168 | |
AY358737.1 | - | 6 - 7 | 881 | 168 | |
BC125167.1 | - | 4 - 5 | 611 | 168 | |
BC125168.1 | - | 4 - 5 | 611 | 168 |