Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLFN5
Entrez Gene ID
162394
Gene Name
schlafen family member 5
Gene Aliases
-
Chromosome Location
Chr.17: 35243026 - 35273650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144975.3 NP_659412.3 3 - 4 1247 111
GenBank mRNA AK054668.1 - 3 - 4 1242 111
BC125200.1 - 2 - 3 1183 111
BC125201.1 - 2 - 3 1183 111
BX647942.1 - 3 - 4 1243 111

View Full Product Details