The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032829.2 | NP_116218.2 | 1 - 2 | 516 | 96 |
GenBank mRNA | BC020193.1 | - | 1 - 2 | 513 | 96 |
DN994281.1 | - | 1 - 2 | 521 | 96 |