Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6V1
Entrez Gene ID
346517
Gene Name
olfactory receptor family 6 subfamily V member 1
Gene Aliases
GPR138
Chromosome Location
Chr.7: 143052341 - 143053282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001667.1 NP_001001667.1 1 - 1 850 79
GenBank mRNA BC137044.1 - 1 - 1 871 79

View Full Product Details