The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253727.1 | NP_001240656.1 | 13 - 14 | 1386 | 161 |
NM_001253728.1 | NP_001240657.1 | 12 - 13 | 1206 | 161 | |
NM_001253732.1 | NP_001240661.1 | 14 - 15 | 1389 | 161 | |
NM_001253733.1 | NP_001240662.1 | 13 - 14 | 1317 | 161 | |
NM_021634.3 | NP_067647.2 | 13 - 14 | 1305 | 161 | |
NR_045579.1 | 13 - 14 | 1922 | 161 | ||
NR_045580.1 | 13 - 14 | 1358 | 161 | ||
NR_045581.1 | 13 - 14 | 1329 | 161 | ||
NR_045584.1 | 14 - 15 | 1358 | 161 | ||
XM_011532175.1 | XP_011530477.1 | 12 - 13 | 1283 | 161 | |
XM_011532176.1 | XP_011530478.1 | 12 - 13 | 1205 | 161 | |
XM_017008519.1 | XP_016864008.1 | 14 - 15 | 1319 | 161 | |
XM_017008520.1 | XP_016864009.1 | 13 - 14 | 1408 | 161 | |
XM_017008521.1 | XP_016864010.1 | 14 - 15 | 1238 | 161 | |
XM_017008526.1 | XP_016864015.1 | 8 - 9 | 776 | 161 | |
GenBank mRNA | AB208943.1 | - | 14 - 15 | 1127 | 161 |
AF190500.1 | - | 13 - 14 | 1042 | 161 | |
AK122647.1 | - | 15 - 16 | 1346 | 161 | |
AK294694.1 | - | 13 - 14 | 1150 | 161 | |
AK295040.1 | - | 13 - 14 | 1745 | 161 | |
AK300379.1 | - | 12 - 13 | 1101 | 161 | |
AK312470.1 | - | 13 - 14 | 1149 | 161 | |
AY899848.1 | - | 14 - 15 | 1095 | 161 | |
AY899850.1 | - | 12 - 13 | 943 | 161 | |
BC112142.1 | - | 13 - 14 | 1124 | 161 | |
BC113617.1 | - | 13 - 14 | 1127 | 161 | |
BC143556.1 | - | 13 - 14 | 1147 | 161 | |
BC143558.1 | - | 13 - 14 | 1180 | 161 | |
BX647975.1 | - | 13 - 14 | 1243 | 161 | |
BX647980.1 | - | 13 - 14 | 1243 | 161 | |
BX647985.1 | - | 13 - 14 | 1272 | 161 |