The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039.3 | NP_001030.2 | 2 - 3 | 456 | 81 |
GenBank mRNA | BC059391.1 | - | 2 - 3 | 456 | 81 |
BC069652.1 | - | 1 - 2 | 317 | 81 | |
DQ402524.1 | - | 1 - 2 | 317 | 81 | |
L36592.1 | - | 1 - 2 | 339 | 81 | |
X87160.1 | - | 1 - 2 | 317 | 81 |