The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039.3 | NP_001030.2 | 4 - 5 | 948 | 64 |
GenBank mRNA | BC059391.1 | - | 4 - 5 | 948 | 64 |
BC069652.1 | - | 3 - 4 | 809 | 64 | |
DQ402524.1 | - | 3 - 4 | 809 | 64 | |
X87160.1 | - | 3 - 4 | 809 | 64 |