The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039.3 | NP_001030.2 | 7 - 8 | 1317 | 76 |
GenBank mRNA | BC059391.1 | - | 7 - 8 | 1317 | 76 |
BC069652.1 | - | 6 - 7 | 1178 | 76 | |
DQ402524.1 | - | 6 - 7 | 1178 | 76 | |
L36592.1 | - | 6 - 7 | 1200 | 76 | |
X87160.1 | - | 6 - 7 | 1178 | 76 |