The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007267.2 | NP_001007268.1 | 12 - 13 | 2243 | 112 |
NM_001195641.1 | NP_001182570.1 | 12 - 13 | 2243 | 112 | |
NM_007366.4 | NP_031392.3 | 12 - 13 | 2243 | 112 | |
XM_005246392.3 | XP_005246449.1 | 12 - 13 | 2131 | 112 | |
XM_011510820.2 | XP_011509122.1 | 12 - 13 | 2131 | 112 | |
XM_017003598.1 | XP_016859087.1 | 12 - 13 | 2131 | 112 | |
XM_017003599.1 | XP_016859088.1 | 12 - 13 | 2131 | 112 | |
XM_017003600.1 | XP_016859089.1 | 2 - 3 | 354 | 112 | |
XM_017003601.1 | XP_016859090.1 | 12 - 13 | 2530 | 112 | |
GenBank mRNA | BC140823.1 | - | 12 - 13 | 2053 | 112 |
BC144631.1 | - | 12 - 13 | 2053 | 112 | |
BF805380.1 | - | 2 - 3 | 304 | 112 | |
D30780.1 | - | 3 - 4 | 438 | 112 | |
U17033.1 | - | 12 - 13 | 2243 | 112 | |
U17034.1 | - | 12 - 13 | 2243 | 112 |