The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007267.2 | NP_001007268.1 | 21 - 22 | 3229 | 71 |
NM_001195641.1 | NP_001182570.1 | 21 - 22 | 3229 | 71 | |
NM_007366.4 | NP_031392.3 | 21 - 22 | 3229 | 71 | |
XM_005246392.3 | XP_005246449.1 | 21 - 22 | 3117 | 71 | |
XM_011510820.2 | XP_011509122.1 | 21 - 22 | 3117 | 71 | |
XM_017003598.1 | XP_016859087.1 | 21 - 22 | 3117 | 71 | |
XM_017003599.1 | XP_016859088.1 | 21 - 22 | 3117 | 71 | |
XM_017003600.1 | XP_016859089.1 | 11 - 12 | 1340 | 71 | |
XM_017003602.1 | XP_016859091.1 | 10 - 11 | 1066 | 71 | |
GenBank mRNA | BC140823.1 | - | 21 - 22 | 3039 | 71 |
BC144631.1 | - | 21 - 22 | 3039 | 71 | |
BF725498.1 | - | 1 - 2 | 96 | 71 | |
U17033.1 | - | 21 - 22 | 3229 | 71 | |
U17034.1 | - | 21 - 22 | 3229 | 71 |